Learn More
Cayman Chemical 4-hydroxy Nonal-d3 50ug
SDP
Supplier:  Cayman Chemical 33210150 UG
Encompass_Preferred
An internal standard for the quantification of 4-HNE by GC- or LC-MS
Catalog No. 50-294-9691