Learn More
Cayman Chemical 4-hydroxy Nonal GlutathIon 1mg
SDP
Catalog No. 502893506
Encompass_Preferred
An internal standard for the quantification of 4-HNE-GSH by GC- or LC-MS
Catalog No. 50-289-3506 Supplier Cayman Chemical Supplier No. 90008761 MG
An internal standard for the quantification of 4-HNE-GSH by GC- or LC-MS