Learn More
Cayman Chemical 4-hydroxy Nonenal, 75899-68-2, 10 mg
SDP
Supplier:  Cayman Chemical 3210010
Encompass_Preferred
Molecular Formula C9H16O2, Purity ≥98%, Formula Weight 156.2, Synonyms: (±)-4-hydroxy-2E-nonenal, 4-HNE; A lipid peroxidation product derived from oxidized -6 PUFAs such as linoleic acid and arachidonic acid which is widely used as a marker of lipid peroxidation.
Catalog No. NC9962070