Learn More
Cayman Chemical 4-hydroxy Nonenal Glutathione
SDP
Supplier:  Cayman Chemical 9000876500 UG
Encompass_Preferred
An internal standard for the quantification of 4-HNE-GSH by GC- or LC-MS
Catalog No. 50-289-4466