Learn More
Cayman Chemical 4-hydroxy Nonenal Glutathione
SDP
Catalog No. 502894465
Encompass_Preferred
An internal standard for the quantification of 4-HNE-GSH by GC- or LC-MS
Catalog No. 50-289-4465 Supplier Cayman Chemical Supplier No. 9000876100 UG
An internal standard for the quantification of 4-HNE-GSH by GC- or LC-MS