Learn More
Cayman Chemical 4-hydroxy Nonenal Mercapturic
SDP
Catalog No. 502895108
Encompass_Preferred
An internal standard for the quantification of 4-HNE mercapturic acid by GC- or LC-MS
Catalog No. 50-289-5108 Supplier Cayman Chemical Supplier No. 9000348100 UG
An internal standard for the quantification of 4-HNE mercapturic acid by GC- or LC-MS