Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxy nonenal mercapturic acid-d3 | 146764-24-1 | MFCD03412011 | 98.0% | 322.44 | C14H22D3NO5S | 1 MG
SDP

Catalog No. 5000284491
Encompass_Preferred

4-Hydroxy nonenal mercapturic acid-d3 is a deuterium-labeled internal standard for analytical quantification of 4-HNE mercapturic acid. It is supplied as an isotopically labeled tracer for use in LC-MS or GC-MS methods.

  • Deuterium-labeled (d3) internal standard for quantitative analysis
  • Molecular formula C14H22D3NO5S and molecular weight 322.44
  • Suitable for LC-MS and GC-MS as a tracer or spike-in standard
  • Purity commonly reported around 98% (see COA for exact value)
  • Refer to the certificate of analysis for storage and handling instructions

Catalog No. 50-002-84491 Supplier Medchemexpress LLC Supplier No. HYW6542141MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxy nonenal mercapturic acid-d3 is a deuterium-labeled internal standard for analytical quantification of 4-HNE mercapturic acid. It is supplied as an isotopically labeled tracer for use in LC-MS or GC-MS methods.

  • Deuterium-labeled (d3) internal standard for quantitative analysis
  • Molecular formula C14H22D3NO5S and molecular weight 322.44
  • Suitable for LC-MS and GC-MS as a tracer or spike-in standard
  • Purity commonly reported around 98% (see COA for exact value)
  • Refer to the certificate of analysis for storage and handling instructions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.