Learn More
Cayman Chemical -4-hydroxy Proprnololhy 10mg
SDP
Catalog No. 502965028
Encompass_Preferred
An active metabolite of (±)-propranolol; decreases isoproterenol-induced tachycardia and vasodepression in cats (EC50 = 54 UG/kg); increases heart rate in catecholamine-depleted rats at 10 and 80 UG/kg; decreases heart rate, myocardial contractility, and cardiac output in dogs in a dose-dependent manner; reduces superoxide-induced lipid peroxidation in isolated rat hepatic microsomes (IC50 = 1.1 µM) and prevents superoxide-induced depletion of GSH levels in BAEC cells (EC50 = 1.2 µM)
Catalog No. 50-296-5028 Supplier Cayman Chemical Supplier No. 1863010 MG
An active metabolite of (±)-propranolol; decreases isoproterenol-induced tachycardia and vasodepression in cats (EC50 = 54 UG/kg); increases heart rate in catecholamine-depleted rats at 10 and 80 UG/kg; decreases heart rate, myocardial contractility, and cardiac output in dogs in a dose-dependent manner; reduces superoxide-induced lipid peroxidation in isolated rat hepatic microsomes (IC50 = 1.1 µM) and prevents superoxide-induced depletion of GSH levels in BAEC cells (EC50 = 1.2 µM)