Learn More
Cayman Chemical (E/Z)-4-hydroxy Tamoxifen, 68392-35-8, 10 mg
SDP
Supplier:  Cayman Chemical 1730810
Encompass_Preferred
Molecular Formula C26H29NO2, Purity ≥98% (E) and (Z) isomers (50:50), Formula Weight 387.5, Synonyms: 4-[1-[4-[2-(dimethylamino)ethoxy]phenyl]-2-phenyl-1-buten-1-yl]-phenol, Afimoxifene, 4-OHT, 4-hydroxy Tamoxifen; An active metabolite of tamoxifen that is formed by the action of CYP2D6 in human liver; inhibits MCF-7 and MDA-MB 231 cell proliferation with IC50 values of 27 and 18 M, respectively; stimulates LC3 lipidation and formation of autophagic vesicles.
Catalog No. NC1213597
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.