Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-TEMPO, free radical, 98+%

Catalog No. AAA1249706
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1249706 5 g
AAA1249703 1 g
AAA1249714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1249706 Supplier Thermo Scientific Chemicals Supplier No. A1249706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2226-96-2 | C9H18NO2

Superoxide scavenger with neuroprotective, anti-inflammatory and analgesic effects4-Hydroxy-TEMPO free radical is used as a catalyst and a chemical oxidant by virtue of being a stable radical. It is used for detoxifying reactive oxygen species in medicinal chemistry. It is involved in the catalysis of disproportionation of superoxide and facilitates hydrogen peroxide metabolism. It is used to prepare tris(2,2,6,6-tetramethyl-1-oxyl-4-piperidyloxy)trimellitate by reacting with benzene-1,2,4-tricarbonyl trichloride. Further, it is used as a free radical scavenger and exhibits neuroprotective, anti-inflammatory and analgesic effects. In addition to this, it has good inhibition effect to acrylic acid esters, methyl acrylate, acrylic acid, acrylonitrile, styrene and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Superoxide scavenger with neuroprotective, anti-inflammatory and analgesic effects4-Hydroxy-TEMPO free radical is used as a catalyst and a chemical oxidant by virtue of being a stable radical. It is used for detoxifying reactive oxygen species in medicinal chemistry. It is involved in the catalysis of disproportionation of superoxide and facilitates hydrogen peroxide metabolism. It is used to prepare tris(2,2,6,6-tetramethyl-1-oxyl-4-piperidyloxy)trimellitate by reacting with benzene-1,2,4-tricarbonyl trichloride. Further, it is used as a free radical scavenger and exhibits neuroprotective, anti-inflammatory and analgesic effects. In addition to this, it has good inhibition effect to acrylic acid esters, methyl acrylate, acrylic acid, acrylonitrile, styrene and butadiene.

Solubility
Soluble in water, ethanol, methanol and dimethyl sulfoxide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2226-96-2
Molecular Formula C9H18NO2
MDL Number MFCD00006478
Synonym 4-Hydroxy-2,2,6,6-tetramethyl-1-piperidin-1-yloxy, free radical

Specifications

Melting Point 69°C to 72°C
pH 8.2
Flash Point 146°C (295°F)
Quantity 5 g
Beilstein 1422990
Solubility Information Soluble in water,ethanol,methanol and dimethyl sulfoxide.
Formula Weight 172.25
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxy-TEMPO, free radical
Hazard Category H302-H318-H373
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
EINECSNumber 218-760-9
RTECSNumber TN8991000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.