Learn More
Medchemexpress LLC 4-hydroxy-TEMPO (standard) | 2226-96-2 | >98.0% | 172.24 g/mol | C9H18NO2 | 25 G
SDP
Supplier:  Medchemexpress LLC HY10056125G
Encompass_Preferred
Tempol (4-hydroxy-TEMPO) is a stable nitroxide radical used as a superoxide dismutase (SOD)-mimetic and reactive oxygen species (ROS) scavenger. It is provided as an analytical standard for research and analytical applications and is commonly used in oxidative stress assays, electron spin resonance (ESR) spin-labeling, and chromatographic or mass-spectrometry methods.
  • Acts as an SOD-mimetic and efficiently scavenges ROS.
  • Stable, cell-permeable nitroxide suitable for biological studies.
  • Supplied as an analytical standard for quantitative and qualitative analysis.
  • Suitable as a spin label for ESR and related spectroscopy techniques.
  • Applicable in oxidative stress assays, HPLC, GC, and MS workflows.
  • Documentation available, including safety data and handling instructions.
Catalog No. 50-002-12711