Learn More
Medchemexpress LLC 4-hydroxyacetophenone | 99-93-4 | MFCD00002359 | >98.0% | 136.15 g/mol | C8H8O2 | 500mg
SDP
Catalog No. 5000268903
Encompass_Preferred
4-Hydroxyacetophenone (P-hydroxyacetophenone) is a major hepatoprotective and choleretic compound found in Artemisia and Illicium plants exhibiting antiviral and anti-inflammatory effects against hepatitis B virus Additionally 4-Hydroxyacetophenone inhibits cancer cell adhesion invasion and migration by remodeling actin 4-Hydroxyacetophenone holds promise for research in the fields of inflammatory diseases and cancer[1][2]
Catalog No. 50-002-68903 Supplier Medchemexpress LLC Supplier No. HYY0073500MG
4-Hydroxyacetophenone (P-hydroxyacetophenone) is a major hepatoprotective and choleretic compound found in Artemisia and Illicium plants exhibiting antiviral and anti-inflammatory effects against hepatitis B virus Additionally 4-Hydroxyacetophenone inhibits cancer cell adhesion invasion and migration by remodeling actin 4-Hydroxyacetophenone holds promise for research in the fields of inflammatory diseases and cancer[1][2]