Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Hydroxyamino)quinoline N-Oxide 97.0+%, TCI America™
SDP

Catalog No. H05155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05155G 5 g
H05151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05155G Supplier TCI America Supplier No. H05155G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4637-56-3
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00059752
InChI Key KCGKBOFRVKLKOO-NTMALXAHSA-N
PubChem CID 5460367
IUPAC Name (NZ)-N-(1-hydroxyquinolin-4-ylidene)hydroxylamine
SMILES C1=CC=C2C(=C1)C(=NO)C=CN2O

Specifications

Melting Point 214°C
Color Yellow
Quantity 5 g
Synonym N-(4-Quinolyl)hydroxylamine 1′-Oxide
Formula Weight 176.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material 4-(Hydroxyamino)quinoline N-Oxide
RTECSNumber VB9800000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.