Learn More
Medchemexpress LLC 4-Hydroxyantipyrine | 1672-63-5 | 99.9% | 100 MG
SDP
Catalog No. 5000410092
Encompass_Preferred
4-Hydroxyantipyrine (4-Hydroxyphenazone; NSC 174055) is the major metabolite of Antipyrine and can function as a biodistribution promoter. It can increase the distribution of the concentration ratio of Citicoline and Antipyrine in the brain.
  • Major metabolite of Antipyrine
  • Can act as a biodistribution promoter
  • Increases distribution of concentration ratio of Citicoline and Antipyrine in the brain
  • Increases the tissue-to-plasma concentration ratio of Citicoline in the brain and liver
  • Increases the tissue-to-plasma concentration ratio of thiopental sodium in the brain, liver, and heart
  • Enhances the blood-brain barrier (BBB) permeability of Antipyrine
Catalog No. 50-004-10092 Supplier Medchemexpress LLC Supplier No. HYB2150100MG
4-Hydroxyantipyrine (4-Hydroxyphenazone; NSC 174055) is the major metabolite of Antipyrine and can function as a biodistribution promoter. It can increase the distribution of the concentration ratio of Citicoline and Antipyrine in the brain.
  • Major metabolite of Antipyrine
  • Can act as a biodistribution promoter
  • Increases distribution of concentration ratio of Citicoline and Antipyrine in the brain
  • Increases the tissue-to-plasma concentration ratio of Citicoline in the brain and liver
  • Increases the tissue-to-plasma concentration ratio of thiopental sodium in the brain, liver, and heart
  • Enhances the blood-brain barrier (BBB) permeability of Antipyrine