Learn More
Medchemexpress LLC 4-Hydroxyantipyrine | 1672-63-5 | 99.9% | 250 MG
SDP
Catalog No. 5000413732
Encompass_Preferred
4-Hydroxyantipyrine is the primary metabolite of Antipyrine and functions as a biodistribution promoter. It has been shown to increase the distribution concentration ratio of Citicoline and Antipyrine in the brain.
  • Enhances tissue-to-plasma concentration ratio of Citicoline in the brain and liver.
  • Enhances tissue-to-plasma concentration ratio of thiopental sodium in the brain, liver, and heart.
  • Improves blood-brain barrier (BBB) permeability of Antipyrine by increasing its Kp value in the brain.
Catalog No. 50-004-13732 Supplier Medchemexpress LLC Supplier No. 250MGHYB2150
4-Hydroxyantipyrine is the primary metabolite of Antipyrine and functions as a biodistribution promoter. It has been shown to increase the distribution concentration ratio of Citicoline and Antipyrine in the brain.
  • Enhances tissue-to-plasma concentration ratio of Citicoline in the brain and liver.
  • Enhances tissue-to-plasma concentration ratio of thiopental sodium in the brain, liver, and heart.
  • Improves blood-brain barrier (BBB) permeability of Antipyrine by increasing its Kp value in the brain.