Learn More
Medchemexpress LLC 4-hydroxybenzaldehyde | 123-08-0 | MFCD00006939 | 100.0% | 122.12 | C7H6O2 | 10g
SDP
Catalog No. 5000269539
Encompass_Preferred
p-Hydroxybenzaldehyde is a one of the major components in vanilla aroma with antagonistic effect on GABAA receptor of the 1 2 2S subtype at high concentrations
Catalog No. 50-002-69539 Supplier Medchemexpress LLC Supplier No. HYY031310G
p-Hydroxybenzaldehyde is a one of the major components in vanilla aroma with antagonistic effect on GABAA receptor of the 1 2 2S subtype at high concentrations