Learn More
Medchemexpress LLC HY-Y0264 500mg Medchemexpress, 4-Hydroxybenzoic acid CAS:99-96-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0264500MG
Encompass_Preferred
Medchemexpress, HY-Y0264 500mg 4-Hydroxybenzoic acid CAS:99-96-7 4-Hydroxybenzoic acid, a phenolic derivative of benzoic acid, could inhibit most gram-positive and some gram-negative bacteria, with an IC50 of 160 μg/mL. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3528