Learn More
Medchemexpress LLC 4-hydroxybenzoic acid | 99-96-7 | MFCD00002547 | 100.0% | 138.12 | C7H6O3 | 1mL
SDP
Catalog No. 5000267317
Encompass_Preferred
4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL
Catalog No. 50-002-67317 Supplier Medchemexpress LLC Supplier No. HYY026410MM/1ML
4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL