Learn More
Medchemexpress LLC 4-hydroxybenzoic acid | 99-96-7 | MFCD00002547 | >98.0% | 138.12 g/mol | C7H6O3 | 100mg
SDP
Catalog No. 5000267354
Encompass_Preferred
4-Hydroxybenzoic acid (Standard) is the analytical standard of 4-Hydroxybenzoic acid This product is intended for research and analytical applications 4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL
Catalog No. 50-002-67354 Supplier Medchemexpress LLC Supplier No. HYY0264R100MG
4-Hydroxybenzoic acid (Standard) is the analytical standard of 4-Hydroxybenzoic acid This product is intended for research and analytical applications 4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL