Learn More
Medchemexpress LLC 4-hydroxybenzoic acid | 99-96-7 | MFCD00002547 | ≥98.0% | 138.12 g/mol | C7H6O3 | 5g
SDP
Catalog No. 5000266986
Encompass_Preferred
4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL
Catalog No. 50-002-66986 Supplier Medchemexpress LLC Supplier No. HYY02645G
4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL