Learn More
Medchemexpress LLC 4-hydroxybenzoic acid-d4 | 152404-47-2 | MFCD01074045 | 100.0% | 142.15 | C7H2D4O3 | 100mg
SDP
Catalog No. 5000267395
Encompass_Preferred
4-Hydroxybenzoic acid-d4 is the deuterium labeled 4-Hydroxybenzoic acid 4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL
Catalog No. 50-002-67395 Supplier Medchemexpress LLC Supplier No. HYY0264S1100MG
4-Hydroxybenzoic acid-d4 is the deuterium labeled 4-Hydroxybenzoic acid 4-Hydroxybenzoic acid a phenolic derivative of benzoic acid could inhibit most gram-positive and some gram-negative bacteria with an IC50 of 160 g/mL