Learn More
Medchemexpress LLC 4-hydroxybenzyl alcohol | 623-05-2 | MFCD00004658 | >98.0% | 124.14 g/mol | C7H8O2 | 250mg
SDP
Catalog No. 5000268466
Encompass_Preferred
4-Hydroxybenzyl alcohol (Standard) is the analytical standard of 4-Hydroxybenzyl alcohol This product is intended for research and analytical applications 4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants Anti-inflammatory anti-oxidant anti-nociceptive activity Neuroprotective effect Inhibitor of tumor angiogenesis and growth[1][2][3][4]
Catalog No. 50-002-68466 Supplier Medchemexpress LLC Supplier No. HYY0892R250MG
4-Hydroxybenzyl alcohol (Standard) is the analytical standard of 4-Hydroxybenzyl alcohol This product is intended for research and analytical applications 4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants Anti-inflammatory anti-oxidant anti-nociceptive activity Neuroprotective effect Inhibitor of tumor angiogenesis and growth[1][2][3][4]