Learn More
Medchemexpress LLC 4-hydroxybenzyl alcohol | 623-05-2 | MFCD00004658 | 99.8% | 124.14 g/mol | C7H8O2 | 50g
SDP
Catalog No. 5000270010
Encompass_Preferred
4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants Anti-inflammatory anti-oxidant anti-nociceptive activity Neuroprotective effect Inhibitor of tumor angiogenesis and growth[1][2][3][4]
Catalog No. 50-002-70010 Supplier Medchemexpress LLC Supplier No. HYY089250G
4-Hydroxybenzyl alcohol is a phenolic compound widely distributed in various kinds of plants Anti-inflammatory anti-oxidant anti-nociceptive activity Neuroprotective effect Inhibitor of tumor angiogenesis and growth[1][2][3][4]