Learn More
Medchemexpress LLC 4-Hydroxybenzylamine | 696-60-6 | 123.16 | 25 G
SDP
Catalog No. 5000448128
Encompass_Preferred
4-Hydroxybenzylamine is a natural biogenic amine that can be produced through the breakdown metabolism of glucosinolates, with glutamine serving as the non-classical ammonia donor. It can be utilized for the study of plant amine biosynthesis.
  • Natural biogenic amine
  • Produced through glucosinolate metabolism
  • Glutamine is a non-classical ammonia donor
  • Useful for plant amine biosynthesis studies
  • Solid form
  • Molecular formula is C7H9NO
  • Classified as phenol, monophenol
  • Endogenous metabolite from plants
  • NMR and MS spectra consistent with structure
Catalog No. 50-004-48128 Supplier Medchemexpress LLC Supplier No. 25GHYW004078
4-Hydroxybenzylamine is a natural biogenic amine that can be produced through the breakdown metabolism of glucosinolates, with glutamine serving as the non-classical ammonia donor. It can be utilized for the study of plant amine biosynthesis.
  • Natural biogenic amine
  • Produced through glucosinolate metabolism
  • Glutamine is a non-classical ammonia donor
  • Useful for plant amine biosynthesis studies
  • Solid form
  • Molecular formula is C7H9NO
  • Classified as phenol, monophenol
  • Endogenous metabolite from plants
  • NMR and MS spectra consistent with structure