Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxybiphenyl-d9 | 126840-28-6 | 179.26 g/mol | C12HD9O | 10mg
SDP

Catalog No. 5000268180
Encompass_Preferred

[1 1 -Biphenyl]-4-ol-d9 is the deuterium labeled [1 1 -Biphenyl]-4-ol[1]

Catalog No. 50-002-68180 Supplier Medchemexpress LLC Supplier No. HYY0162S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[1 1 -Biphenyl]-4-ol-d9 is the deuterium labeled [1 1 -Biphenyl]-4-ol[1]

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.