Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxycarbazole | 52602-39-8 | MFCD02178385 | 99.3% | 183.21 g/mol | C12H9NO | 25 G
SDP

Catalog No. 5000302759
Encompass_Preferred

9H-Carbazol-4-ol (4-hydroxycarbazole) is an aromatic heterocyclic compound used in organic synthesis and research applications. It is supplied as a solid with reported analytical purity and standard storage instructions for laboratory use.

  • High reported purity: 99.29%.
  • Solid form, off-white to light brown appearance.
  • Molecular formula C12H9NO and molecular weight 183.21 g/mol.
  • Packaged for laboratory use in 25 g quantities.
  • Storage: powder stable at -20°C for up to 3 years; SDS available for handling.

Catalog No. 50-003-02759 Supplier Medchemexpress LLC Supplier No. 25GHYW016331
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

9H-Carbazol-4-ol (4-hydroxycarbazole) is an aromatic heterocyclic compound used in organic synthesis and research applications. It is supplied as a solid with reported analytical purity and standard storage instructions for laboratory use.

  • High reported purity: 99.29%.
  • Solid form, off-white to light brown appearance.
  • Molecular formula C12H9NO and molecular weight 183.21 g/mol.
  • Packaged for laboratory use in 25 g quantities.
  • Storage: powder stable at -20°C for up to 3 years; SDS available for handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.