Learn More
Selleck Chemical LLC 4-Hydroxychalcone S4937-100mg
SDP
Catalog No. 503109082
Encompass_Preferred
4 -Hydroxychalcone (P-Cinnamoylphenol) found in herbs and spices and tea is a member of the class of compounds known as retrochalcones It has diverse biological activities inhibiting TNF -induced NF- B pathway activation in a dose-dependent manner and activating BMP signaling
Catalog No. 50-310-9082 Supplier Selleck Chemical LLC Supplier No. S4937100MG
4 -Hydroxychalcone (P-Cinnamoylphenol) found in herbs and spices and tea is a member of the class of compounds known as retrochalcones It has diverse biological activities inhibiting TNF -induced NF- B pathway activation in a dose-dependent manner and activating BMP signaling