Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxychalcone | 20426-12-4 | MFCD00016488 | 99.3% | 224.25 g/mol | C15H12O2 | 1 ML
SDP

Catalog No. 5000240276
Encompass_Preferred

4-Hydroxychalcone is a chalcone metabolite supplied as a 10 mM solution in DMSO for research use. It is reported to inhibit TNF-α-induced NF-κB activation and to exhibit anti-angiogenic and anti-inflammatory effects, providing a high-purity, ready-to-use stock for cell-based and biochemical assays. Handle under recommended storage conditions and protect from light.

  • 10 mM solution in DMSO.
  • High purity: 99.3%.
  • Molecular formula C15H12O2; molecular weight 224.25 g/mol.
  • CAS number 20426-12-4.
  • Inhibits TNF-α-induced NF-κB pathway; anti-angiogenic and anti-inflammatory activity.
  • Store protected from light; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Catalog No. 50-002-40276 Supplier Medchemexpress LLC Supplier No. HY10781810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Hydroxychalcone is a chalcone metabolite supplied as a 10 mM solution in DMSO for research use. It is reported to inhibit TNF-α-induced NF-κB activation and to exhibit anti-angiogenic and anti-inflammatory effects, providing a high-purity, ready-to-use stock for cell-based and biochemical assays. Handle under recommended storage conditions and protect from light.

  • 10 mM solution in DMSO.
  • High purity: 99.3%.
  • Molecular formula C15H12O2; molecular weight 224.25 g/mol.
  • CAS number 20426-12-4.
  • Inhibits TNF-α-induced NF-κB pathway; anti-angiogenic and anti-inflammatory activity.
  • Store protected from light; in solvent: -80°C up to 6 months, -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.