Learn More
TARGETMOL CHEMICALS INC 4-Hydroxychalcone 500MG
SDP
Catalog No. 502876982
Encompass_Preferred
Also available in 5 g, 1 mL, 100 mg and bulk. Please contact Fisher for quotes. 4'-Hydroxychalcone (2-Benzal-4-Hydroxyacetophenone) is a chalcone metabolite with hepatoprotective activity Purity 99.86%
Catalog No. 50-287-6982 Supplier TARGETMOL CHEMICALS INC Supplier No. T7917500MG
Also available in 5 g, 1 mL, 100 mg and bulk. Please contact Fisher for quotes. 4'-Hydroxychalcone (2-Benzal-4-Hydroxyacetophenone) is a chalcone metabolite with hepatoprotective activity Purity 99.86%