Learn More
Medchemexpress LLC 4-Hydroxycoumarin | 1076-38-6 | 100 G
SDP
Catalog No. 5000419488
Encompass_Preferred
4-Hydroxycoumarin is an orally active coumarin derivative, serving as a versatile heterocyclic scaffold for organic compound synthesis. It exhibits both electrophilic and nucleophilic properties. This compound acts as an HIV protease and tyrosine kinase inhibitor, demonstrating anti-inflammatory, antibacterial, and anti-tumor effects. It is primarily utilized as a laboratory chemical and in the manufacturing of various substances.
  • Orally active coumarin derivative
  • Versatile scaffold for organic synthesis
  • Inhibits HIV protease and tyrosine kinase
  • Exhibits anti-inflammatory, antibacterial, and anti-tumor effects
  • Primarily used as a laboratory chemical
  • Solid physical state, off-white appearance
  • Chemically stable under normal conditions
Catalog No. 50-004-19488 Supplier Medchemexpress LLC Supplier No. HYN6856100G
4-Hydroxycoumarin is an orally active coumarin derivative, serving as a versatile heterocyclic scaffold for organic compound synthesis. It exhibits both electrophilic and nucleophilic properties. This compound acts as an HIV protease and tyrosine kinase inhibitor, demonstrating anti-inflammatory, antibacterial, and anti-tumor effects. It is primarily utilized as a laboratory chemical and in the manufacturing of various substances.
  • Orally active coumarin derivative
  • Versatile scaffold for organic synthesis
  • Inhibits HIV protease and tyrosine kinase
  • Exhibits anti-inflammatory, antibacterial, and anti-tumor effects
  • Primarily used as a laboratory chemical
  • Solid physical state, off-white appearance
  • Chemically stable under normal conditions