Learn More
Medchemexpress LLC 4-Hydroxycoumarin | 1076-38-6 | 250 G
SDP
Catalog No. 5000422070
Encompass_Preferred
4-Hydroxycoumarin is an orally active coumarin derivative and a versatile heterocyclic scaffold used in the synthesis of various organic compounds. It exhibits both electrophilic and nucleophilic properties. It is identified as an HIV protease inhibitor and tyrosine kinase inhibitor, and also demonstrates anti-inflammatory, antibacterial, and anti-tumor effects.
  • Acts as an HIV protease inhibitor
  • Acts as a tyrosine kinase inhibitor
  • Exhibits anti-inflammatory effects
  • Exhibits antibacterial effects
  • Exhibits anti-tumor effects
  • Disrupts actin cytoskeleton in B16-F10 melanoma cells
  • Reduces adhesion and motility to extracellular matrix proteins
  • Inhibits tumor growth and improves survival rate in melanoma mice
  • Reduces symptoms in a rat model of trinitrobenzene sulfonic acid-induced colitis
Catalog No. 50-004-22070 Supplier Medchemexpress LLC Supplier No. 250GHYN6856
4-Hydroxycoumarin is an orally active coumarin derivative and a versatile heterocyclic scaffold used in the synthesis of various organic compounds. It exhibits both electrophilic and nucleophilic properties. It is identified as an HIV protease inhibitor and tyrosine kinase inhibitor, and also demonstrates anti-inflammatory, antibacterial, and anti-tumor effects.
  • Acts as an HIV protease inhibitor
  • Acts as a tyrosine kinase inhibitor
  • Exhibits anti-inflammatory effects
  • Exhibits antibacterial effects
  • Exhibits anti-tumor effects
  • Disrupts actin cytoskeleton in B16-F10 melanoma cells
  • Reduces adhesion and motility to extracellular matrix proteins
  • Inhibits tumor growth and improves survival rate in melanoma mice
  • Reduces symptoms in a rat model of trinitrobenzene sulfonic acid-induced colitis