Learn More
Medchemexpress LLC 4-Hydroxycoumarin | 1076-38-6 | 50 G
SDP
Catalog No. 5000419490
Encompass_Preferred
4-Hydroxycoumarin is an orally active coumarin derivative, a versatile heterocyclic scaffold used in synthesizing various organic compounds. It possesses both electrophilic and nucleophilic properties. It acts as an HIV protease inhibitor and tyrosine kinase inhibitor, and exhibits anti-inflammatory, antibacterial, and anti-tumor effects.
  • Orally active coumarin derivative
  • Versatile heterocyclic scaffold for organic compound synthesis
  • Possesses both electrophilic and nucleophilic properties
  • Acts as an HIV protease inhibitor
  • Acts as a tyrosine kinase inhibitor
  • Exhibits anti-inflammatory effects
  • Exhibits antibacterial effects
  • Exhibits anti-tumor effects
Catalog No. 50-004-19490 Supplier Medchemexpress LLC Supplier No. HYN685650G
4-Hydroxycoumarin is an orally active coumarin derivative, a versatile heterocyclic scaffold used in synthesizing various organic compounds. It possesses both electrophilic and nucleophilic properties. It acts as an HIV protease inhibitor and tyrosine kinase inhibitor, and exhibits anti-inflammatory, antibacterial, and anti-tumor effects.
  • Orally active coumarin derivative
  • Versatile heterocyclic scaffold for organic compound synthesis
  • Possesses both electrophilic and nucleophilic properties
  • Acts as an HIV protease inhibitor
  • Acts as a tyrosine kinase inhibitor
  • Exhibits anti-inflammatory effects
  • Exhibits antibacterial effects
  • Exhibits anti-tumor effects