Learn More
Cayman Chemical 4-Hydroxycoumarin, Purity: ≥98%, CAS: 1076-38-6, 250 g
SDP
Catalog No. 502946901
Encompass_Preferred
A coumarin; inhibits vitamin K1 epoxidation and prothrombin synthesis by 18 and 67%, respectively, in cell-free assays at 6.2 mM; has been used in the synthesis of anticoagulants, as well as antithrombotic, anti-inflammatory, antibacterial, and antifungal agents
Catalog No. 50-294-6901 Supplier Cayman Chemical Supplier No. 33568250 G
A coumarin; inhibits vitamin K1 epoxidation and prothrombin synthesis by 18 and 67%, respectively, in cell-free assays at 6.2 mM; has been used in the synthesis of anticoagulants, as well as antithrombotic, anti-inflammatory, antibacterial, and antifungal agents