Learn More
Medchemexpress LLC 4-Hydroxycoumarin (standard) | 1076-38-6 | 50 MG
SDP
Catalog No. 5000419475
Encompass_Preferred
4-Hydroxycoumarin (Standard) is the analytical standard of 4-Hydroxycoumarin, intended for research and analytical applications. It is an orally active coumarin derivative and a versatile heterocyclic scaffold, frequently used in the synthesis of diverse organic compounds. The product exhibits both electrophilic and nucleophilic properties.
  • Analytical standard.
  • Intended for research and analytical applications.
  • Orally active coumarin derivative.
  • Versatile heterocyclic scaffold.
  • Used in the synthesis of various organic compounds.
  • Possesses both electrophilic and nucleophilic properties.
  • Functions as an HIV protease inhibitor.
  • Functions as a tyrosine kinase inhibitor.
  • Displays anti-inflammatory effects.
  • Displays antibacterial effects.
  • Displays anti-tumor effects.
Catalog No. 50-004-19475 Supplier Medchemexpress LLC Supplier No. HYN6856R50MG
4-Hydroxycoumarin (Standard) is the analytical standard of 4-Hydroxycoumarin, intended for research and analytical applications. It is an orally active coumarin derivative and a versatile heterocyclic scaffold, frequently used in the synthesis of diverse organic compounds. The product exhibits both electrophilic and nucleophilic properties.
  • Analytical standard.
  • Intended for research and analytical applications.
  • Orally active coumarin derivative.
  • Versatile heterocyclic scaffold.
  • Used in the synthesis of various organic compounds.
  • Possesses both electrophilic and nucleophilic properties.
  • Functions as an HIV protease inhibitor.
  • Functions as a tyrosine kinase inhibitor.
  • Displays anti-inflammatory effects.
  • Displays antibacterial effects.
  • Displays anti-tumor effects.