Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-hydroxycyclohexanecarboxylic acid | 17419-81-7 | MFCD00077974 | ≥95.0% | 144.17 g/mol | C7H12O3 | 1 ML
SDP

Catalog No. 5000300322
Encompass_Preferred

4-Hydroxycyclohexanecarboxylic acid is an endogenous metabolite in the cyclohexanol family used as a research reagent. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO for analytical and biochemical studies; intended for research use only.

  • ≥95.0% purity.
  • Molecular weight 144.17 g/mol.
  • Chemical formula C7H12O3.
  • Available as solid and 10 mM solution in DMSO.
  • Storage: powder at -20°C; in solvent at -80°C for long-term.
  • Suitable for metabolite analysis and biochemical assays.

Catalog No. 50-003-00322 Supplier Medchemexpress LLC Supplier No. HYW01567510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Hydroxycyclohexanecarboxylic acid is an endogenous metabolite in the cyclohexanol family used as a research reagent. It is offered as a solid and as a ready-to-use 10 mM solution in DMSO for analytical and biochemical studies; intended for research use only.

  • ≥95.0% purity.
  • Molecular weight 144.17 g/mol.
  • Chemical formula C7H12O3.
  • Available as solid and 10 mM solution in DMSO.
  • Storage: powder at -20°C; in solvent at -80°C for long-term.
  • Suitable for metabolite analysis and biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.