Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. H096025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H096025G 25 g
H0960250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H096025G Supplier TCI America Supplier No. H096025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 17419-81-7
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00077974
InChI Key HCFRWBBJISAZNK-UHFFFAOYSA-N
PubChem CID 151138
IUPAC Name 4-hydroxycyclohexane-1-carboxylic acid
SMILES C1CC(CCC1C(=O)O)O

Specifications

Color White
Quantity 25 g
Formula Weight 144.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Hydroxycyclohexanecarboxylic Acid (cis- and trans- mixture)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.