Learn More
Medchemexpress LLC 4-hydroxydiphenylamine (p-anilinophenol) | 122-37-2 | MFCD00020142 | 98.0% | 185.22 g/mol | C12H11NO | 25 G
SDP
Catalog No. 5000209074
Encompass_Preferred
4-Hydroxydiphenylamine (p-anilinophenol) is an aromatic amine used as a research chemical, antioxidant, and intermediate in organic synthesis. It is supplied as solid and solution formats for laboratory research.
  • Aromatic amine used as antioxidant and synthetic intermediate.
  • Reported purity of 98.0%.
  • Molecular weight 185.22 g/mol; formula C12H11NO.
  • Available as solid packages (5 g, 10 g, 25 g, 50 g) and as a 10 mM solution in DMSO.
  • Common applications include environmental science and toxicology research.
Catalog No. 50-002-09074 Supplier Medchemexpress LLC Supplier No. 25GHYW088320
Add to Cart
Add to Cart
4-Hydroxydiphenylamine (p-anilinophenol) is an aromatic amine used as a research chemical, antioxidant, and intermediate in organic synthesis. It is supplied as solid and solution formats for laboratory research.
  • Aromatic amine used as antioxidant and synthetic intermediate.
  • Reported purity of 98.0%.
  • Molecular weight 185.22 g/mol; formula C12H11NO.
  • Available as solid packages (5 g, 10 g, 25 g, 50 g) and as a 10 mM solution in DMSO.
  • Common applications include environmental science and toxicology research.