Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4'-Hydroxyflavanone | 6515-37-3 | 1 ML
SDP

Catalog No. 5000418507
Encompass_Preferred

4'-Hydroxyflavanone is an inhibitor of SREBP maturation and lipid synthesis, and a synthetic analogue of flavanone. It is used in research into hepatic steatosis and dyslipidemia.

  • Purity of 99.75%
  • Molecular weight of 240.25 g/mol
  • Chemical formula C15H12O3
  • Classified as a flavonoid, flavanone, phenol, and monophenol
  • Isolated from Aspergillus niger
  • Provided as a 10 mM solution in DMSO
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store solution at -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-004-18507 Supplier Medchemexpress LLC Supplier No. HYN849510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4'-Hydroxyflavanone is an inhibitor of SREBP maturation and lipid synthesis, and a synthetic analogue of flavanone. It is used in research into hepatic steatosis and dyslipidemia.

  • Purity of 99.75%
  • Molecular weight of 240.25 g/mol
  • Chemical formula C15H12O3
  • Classified as a flavonoid, flavanone, phenol, and monophenol
  • Isolated from Aspergillus niger
  • Provided as a 10 mM solution in DMSO
  • Store powder at -20°C for 3 years, or 4°C for 2 years
  • Store solution at -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.