Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4'-Hydroxyflavanone | 6515-37-3 | 100 MG
SDP

Catalog No. 5000417416
Encompass_Preferred

4'-Hydroxyflavanone is an inhibitor of SREBP maturation and lipid synthesis. It is a synthetic analogue of flavanone and has potential for research into hepatic steatosis and dyslipidemia.

  • Inhibitor of SREBP maturation
  • Inhibitor of lipid synthesis
  • Synthetic analogue of flavanone
  • Potential for hepatic steatosis research
  • Potential for dyslipidemia research
  • Molecular weight: 240.25
  • Formula: C15H12O3
  • Appearance: white to light yellow solid
  • Purity: 99.75%
  • Soluble in DMSO (100 mg/mL)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-17416 Supplier Medchemexpress LLC Supplier No. HYN8495100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4'-Hydroxyflavanone is an inhibitor of SREBP maturation and lipid synthesis. It is a synthetic analogue of flavanone and has potential for research into hepatic steatosis and dyslipidemia.

  • Inhibitor of SREBP maturation
  • Inhibitor of lipid synthesis
  • Synthetic analogue of flavanone
  • Potential for hepatic steatosis research
  • Potential for dyslipidemia research
  • Molecular weight: 240.25
  • Formula: C15H12O3
  • Appearance: white to light yellow solid
  • Purity: 99.75%
  • Soluble in DMSO (100 mg/mL)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.