Learn More
Medchemexpress LLC 4'-Hydroxyflavanone | 6515-37-3 | 50 MG
SDP
Catalog No. 5000417342
Encompass_Preferred
4'-Hydroxyflavanone is an inhibitor of SREBP maturation and lipid synthesis. It is a synthetic analogue of flavanone, with potential for hepatic steatosis and dyslipidemia research.
  • Inhibitor of SREBP maturation
  • Inhibitor of lipid synthesis
  • Synthetic analogue of flavanone
  • Potential for hepatic steatosis research
  • Potential for dyslipidemia research
Catalog No. 50-004-17342 Supplier Medchemexpress LLC Supplier No. HYN849550MG
4'-Hydroxyflavanone is an inhibitor of SREBP maturation and lipid synthesis. It is a synthetic analogue of flavanone, with potential for hepatic steatosis and dyslipidemia research.
  • Inhibitor of SREBP maturation
  • Inhibitor of lipid synthesis
  • Synthetic analogue of flavanone
  • Potential for hepatic steatosis research
  • Potential for dyslipidemia research