Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hydroxyflavanone 98.0+%, TCI America™
SDP

Catalog No. H10261G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H10261G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H10261G Supplier TCI America Supplier No. H10261G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6515-37-3
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.26
MDL Number MFCD00017705
InChI Key ZLHVIYHWWQYJID-UHFFFAOYNA-N
Synonym 4'-hydroxyflavanone, 2-4-hydroxyphenyl chroman-4-one, 4'-hydroxy flavanone, 2,3-dihydro-2-4-hydroxyphenyl-4h-1-benzopyran-4-one, 2-4-hydroxyphenyl-2,3-dihydro-4h-chromen-4-one, chembl73933, 4h-1-benzopyran-4-one, 2,3-dihydro-2-4-hydroxyphenyl, flavanone, 4'-hydroxy, 4-hydroxyflavanone, 4'-hydroxyflavaone
PubChem CID 165506
ChEBI CHEBI:34361
IUPAC Name 2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
SMILES OC1=CC=C(C=C1)C1CC(=O)C2=CC=CC=C2O1

Specifications

Melting Point 192°C
Quantity 1 g
Formula Weight 240.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyflavanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.