Learn More
TARGETMOL CHEMICALS INC 4-Hydroxyflavone 10MG
SDP
Catalog No. 502876712
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 4'-Hydroxyflavone is a flavonoid compound isolated from the roots of Scutellaria baicalensis with anti-inflammatory activity. Purity 99.42%
Catalog No. 50-287-6712 Supplier TARGETMOL CHEMICALS INC Supplier No. T8429610MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 4'-Hydroxyflavone is a flavonoid compound isolated from the roots of Scutellaria baicalensis with anti-inflammatory activity. Purity 99.42%