Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4'-Hydroxyflavone | 4143-63-9 | MFCD00017687 | 97.0% | 238.24 g/mol | C15H10O3 | 10 MG
SDP

Catalog No. 5000281868
Encompass_Preferred

4'-Hydroxyflavone is a flavone-class small molecule used in research as an aldose reductase inhibitor. It is supplied as a high-purity research reagent for biochemical assays and is commonly used in enzyme inhibition studies, structure-activity relationship (SAR) investigations, and small-molecule screening.

  • High purity (97.0%)
  • Molecular formula C15H10O3
  • Molecular weight 238.24 g/mol
  • CAS number 4143-63-9
  • Reported aldose reductase IC50 12 μM
  • Suitable for enzyme inhibition assays and SAR studies

Catalog No. 50-002-81868 Supplier Medchemexpress LLC Supplier No. HYW31909410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4'-Hydroxyflavone is a flavone-class small molecule used in research as an aldose reductase inhibitor. It is supplied as a high-purity research reagent for biochemical assays and is commonly used in enzyme inhibition studies, structure-activity relationship (SAR) investigations, and small-molecule screening.

  • High purity (97.0%)
  • Molecular formula C15H10O3
  • Molecular weight 238.24 g/mol
  • CAS number 4143-63-9
  • Reported aldose reductase IC50 12 μM
  • Suitable for enzyme inhibition assays and SAR studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.