Learn More
Medchemexpress LLC 4'-hydroxyflavone | 4143-63-9 | MFCD00017687 | 97.0% | 238.24 g/mol | C15H10O3 | 5 MG
SDP
Catalog No. 5000282173
Encompass_Preferred
4'-Hydroxyflavone is a small-molecule flavone provided for research use. It has the chemical formula C15H10O3, a molecular weight of 238.24 g/mol, and is reported to inhibit aldose reductase (IC50 = 12 μM). The material is intended as a reference reagent for biochemical, pharmacological, and enzymology studies.
  • High purity (97.0%) suitable for analytical and biological assays.
  • Reported aldose reductase inhibitor (IC50 = 12 μM) for target validation studies.
  • Well-characterized chemical identity and molecular properties.
  • Available in small pack sizes appropriate for research applications.
  • Suitable for biochemical assays, enzymology, and screening workflows.
Catalog No. 50-002-82173 Supplier Medchemexpress LLC Supplier No. HYW3190945MG
4'-Hydroxyflavone is a small-molecule flavone provided for research use. It has the chemical formula C15H10O3, a molecular weight of 238.24 g/mol, and is reported to inhibit aldose reductase (IC50 = 12 μM). The material is intended as a reference reagent for biochemical, pharmacological, and enzymology studies.
  • High purity (97.0%) suitable for analytical and biological assays.
  • Reported aldose reductase inhibitor (IC50 = 12 μM) for target validation studies.
  • Well-characterized chemical identity and molecular properties.
  • Available in small pack sizes appropriate for research applications.
  • Suitable for biochemical assays, enzymology, and screening workflows.