Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hydroxyhexanophenone 99.0+%, TCI America™
SDP

Catalog No. H08845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08845G 5 g
H088425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08845G Supplier TCI America Supplier No. H08845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2589-72-2
Molecular Formula C12H16O2
Molecular Weight (g/mol) 192.258
MDL Number MFCD00671540
InChI Key YDGNMJZDWLQUTE-UHFFFAOYSA-N
PubChem CID 345110
IUPAC Name 1-(4-hydroxyphenyl)hexan-1-one
SMILES CCCCCC(=O)C1=CC=C(C=C1)O

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 5 g
Synonym Amyl 4-Hydroxyphenyl Ketone, 4′C-Hydroxycaprophenone
Formula Weight 192.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxyhexanophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.