Learn More
Medchemexpress LLC 4-Hydroxyhippuric acid | 2482-25-9 | 99.59% | 195.17 | 10 MG
SDP
Catalog No. 5000435957
Encompass_Preferred
4-Hydroxyhippuric acid (Standard) is the analytical standard of 4-Hydroxyhippuric acid. This product is intended for research and analytical applications. 4-Hydroxyhippuric acid is a by-product of metabolism of polyphenols and increases the microflora activity related to the refeeding.
  • The compound is an analytical grade reference standard.
  • It is used in qualitative, quantitative, and methodological research experiments.
  • Commonly applied in techniques such as HPLC, GC, and MS.
Catalog No. 50-004-35957 Supplier Medchemexpress LLC Supplier No. HY113158R10MG
4-Hydroxyhippuric acid (Standard) is the analytical standard of 4-Hydroxyhippuric acid. This product is intended for research and analytical applications. 4-Hydroxyhippuric acid is a by-product of metabolism of polyphenols and increases the microflora activity related to the refeeding.
  • The compound is an analytical grade reference standard.
  • It is used in qualitative, quantitative, and methodological research experiments.
  • Commonly applied in techniques such as HPLC, GC, and MS.