Learn More
Medchemexpress LLC 4-Hydroxyhippuric acid | 2482-25-9 | 99.59% | 195.17 | 50 MG
SDP
Catalog No. 5000437406
Encompass_Preferred
4-Hydroxyhippuric acid (Standard) is the analytical standard of 4-Hydroxyhippuric acid. This product is intended for research and analytical applications. It is a by-product of metabolism of polyphenols and increases microflora activity related to refeeding.
  • Analytical standard for research and analytical applications.
  • Reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-37406 Supplier Medchemexpress LLC Supplier No. HY11315850MG
4-Hydroxyhippuric acid (Standard) is the analytical standard of 4-Hydroxyhippuric acid. This product is intended for research and analytical applications. It is a by-product of metabolism of polyphenols and increases microflora activity related to refeeding.
  • Analytical standard for research and analytical applications.
  • Reference standard supplied assay.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.