Learn More
Medchemexpress LLC 4-Hydroxyhippuric acid | 2482-25-9 | 99.6% | 195.17 | 25 MG
SDP
Catalog No. 5000439410
Encompass_Preferred
4-Hydroxyhippuric acid (Standard) is an analytical standard used for research and analytical applications. It is a by-product of metabolism of polyphenols and increases microflora activity related to refeeding.
  • Analytical standard for research and analytical applications.
  • Reference standard supplied assay.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS.
Catalog No. 50-004-39410 Supplier Medchemexpress LLC Supplier No. 25MGHY113158R
4-Hydroxyhippuric acid (Standard) is an analytical standard used for research and analytical applications. It is a by-product of metabolism of polyphenols and increases microflora activity related to refeeding.
  • Analytical standard for research and analytical applications.
  • Reference standard supplied assay.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS.