Learn More
Medchemexpress LLC 4-Hydroxyhippuric acid | 2482-25-9 | 99.6% | 195.17 | 50 MG
SDP
Catalog No. 5000435675
Encompass_Preferred
4-Hydroxyhippuric acid (Standard) is the analytical standard of 4-Hydroxyhippuric acid. This product is intended for research and analytical applications, serving as a reference for assays. It is a by-product of the metabolism of polyphenols and increases microflora activity related to refeeding, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Reference for various assays
  • By-product of polyphenol metabolism
  • Increases microflora activity related to refeeding
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-004-35675 Supplier Medchemexpress LLC Supplier No. HY113158R50MG
4-Hydroxyhippuric acid (Standard) is the analytical standard of 4-Hydroxyhippuric acid. This product is intended for research and analytical applications, serving as a reference for assays. It is a by-product of the metabolism of polyphenols and increases microflora activity related to refeeding, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Reference for various assays
  • By-product of polyphenol metabolism
  • Increases microflora activity related to refeeding
  • Suitable for HPLC, GC, and MS experiments