Learn More
Cayman Chemical 4-Hydroxyhippuric Acid, Purity: ≥98%, CAS: 2482-25-9, 500 mg
SDP
Catalog No. 502989025
Encompass_Preferred
An OAT1 inhibitor (IC50 = 25 µM in opossum kidney cells); increases the production of free radicals in opossum kidney cells expressing human OAT1; reduces the secretion of TNF-α by 30.4% at 1 µM
Catalog No. 50-298-9025 Supplier Cayman Chemical Supplier No. 30115500 MG
An OAT1 inhibitor (IC50 = 25 µM in opossum kidney cells); increases the production of free radicals in opossum kidney cells expressing human OAT1; reduces the secretion of TNF-α by 30.4% at 1 µM